missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane Dihydrochloride (Low water content), TRC

Product Code. 30367511
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 30367511

Brand: Toronto Research Chemicals TRCD4176955G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Diaminopropane Dihydrochloride
CAS 10517-44-9
Molecular Formula C3H10N2 • 2HCl
Quantity 5 g
Synonym Trimethylenediamine Dihydrochloride
InChI Formula InChI=1S/C3H10N2.2ClH/c4-2-1-3-5;;/h1-5H2;2*1H
SMILES NCCCN.Cl.Cl
IUPAC Name propane-1,3-diamine hydrochloride
Molecular Weight (g/mol) 147.04
Formula Weight 146.04
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.