missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane, 98%

Product Code. 11474230
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474230 100 mL 100mL
11483914 500 mL 500mL
11493914 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11474230 Supplier Thermo Scientific Alfa Aesar Supplier No. A11932.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 109-76-2 | C3H10N2 | 74.127 g/mol

1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

Solubility
Miscible with water.

Notes
Hygroscopic. Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-76-2
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.127
MDL Number MFCD00008228
InChI Key XFNJVJPLKCPIBV-UHFFFAOYSA-N
Synonym 1,3-diaminopropane, 1,3-propanediamine, trimethylenediamine, 1,3-propylenediamine, propandiamine, alpha,omega-propanediamine, unii-cb3isl56kg, ccris 4054, 1,3-propane diamine, cb3isl56kg
PubChem CID 428
ChEBI CHEBI:15725
IUPAC Name propane-1,3-diamine
SMILES C(CN)CN

Specifications

Melting Point -12°C
Density 0.888
pH >12
Boiling Point 134°C to 136°C
Flash Point 48°C (118°F)
Odor Amine-like
Refractive Index 1.4573
Quantity 100 mL
UN Number UN2734
Beilstein 605277
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 74.13
Percent Purity 98%
Chemical Name or Material 1,3-Diaminopropane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Diaminopropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Propanediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.