missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminocyclohexane, 95+%, mixture of cis and trans isomers

Product Code. 10684363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10684363 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10684363 Supplier Thermo Scientific Acros Supplier No. 370240100
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3385-21-5 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3385-21-5
CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Density 0.9500g/mL
Infrared Spectrum Authentic
Assay Percent Range 95% min. (sum of cis and trans) (GC)
Packaging Glass bottle
Molecular Formula C6H14N2
Linear Formula C6H10(NH2)2
Refractive Index 1.49
MDL Number MFCD00059563
Quantity 10 mL
Specific Gravity 0.95
Synonym 1,3-diaminocyclohexane, 1,3-cyclohexanediamine, cyclohex-1,3-ylenediamine, ccris 6679, 2,4-diaminocyclohexane, acmc-20ap4j, 1,3-diamino-cyclohexane, cyclohexane-1.3-diamine, rac-1,3-cyclohexanediamine, 1r,3s-3-azaniumylcyclohexyl azanium
InChI Key GEQHKFFSPGPGLN-UHFFFAOYSA-N
SMILES C1CC(CC(C1)N)N
IUPAC Name cyclohexane-1,3-diamine
Molecular Weight (g/mol) 114.19
PubChem CID 18814
Formula Weight 114.19
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material 1, 3-Diaminocyclohexane, Mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.