missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diamino-2-propanol, 97%

Product Code. 11474167
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474167 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474167 Supplier Thermo Scientific Alfa Aesar Supplier No. A19618.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 616-29-5 | C3H10N2O | 90.13 g/mol

It finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Very soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Specifications

CAS 616-29-5
Melting Point 40°C to 45°C
Boiling Point 235°C
Flash Point >110°C
Molecular Formula C3H10N2O
MDL Number MFCD00008142
Quantity 250 g
Beilstein 741859
Synonym 1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino
Solubility Information Very soluble aq. soln..
InChI Key UYBWIEGTWASWSR-UHFFFAOYSA-N
SMILES NCC(O)CN
IUPAC Name 1,3-diaminopropan-2-ol
Molecular Weight (g/mol) 90.13
PubChem CID 61157
Formula Weight 90.13
Percent Purity 97%
Chemical Name or Material 1,3-Diamino-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.