missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diacetoxybenzene, 97%

Product Code. 11494573
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494573 250 g 250g
11484573 50 g 50g
11474573 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494573 Supplier Thermo Scientific Alfa Aesar Supplier No. A11547.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-58-7 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-58-7
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00008701
InChI Key STOUHHBZBQBYHH-UHFFFAOYSA-N
Synonym 1,3-diacetoxybenzene, resorcinol diacetate, 1,3-benzenediol, diacetate, m-phenylenediacetate, resorcinol, diacetate, m-phenylene di acetate, 1,3-dihydroxybenzene diacetate, dihydroxybenzene diacetate, 1,3-phenylene diacetate, 1,3-benzenediol, 1,3-diacetate
PubChem CID 7942
IUPAC Name (3-acetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC(=CC=C1)OC(=O)C

Specifications

Density 1.179
Boiling Point 278°C
Flash Point >110°C (230°F)
Refractive Index 1.503
Quantity 250 g
Beilstein 1875007
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material 1,3-Diacetoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.