missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diacetin (Technical grade, ~50%), TRC

Product Code. 30357477
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357477 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357477 Supplier Toronto Research Chemicals Supplier No. TRCD304600250G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Diacetin (Technical grade)
CAS 105-70-4
Molecular Formula C7H12O5
Quantity 250 g
Synonym 1,3-Diacetoxy-2-propanol, Glycerine 1,3-Diacetate, Glycerol 1,3-Diacetate, 1,2,3-Propanetriol 1,3-Diacetate
InChI Formula InChI=1S/C7H12O5/c1-5(8)11-3-7(10)4-12-6(2)9/h7,10H,3-4H2,1-2H3
SMILES CC(=O)OCC(O)COC(=O)C
IUPAC Name (3-acetyloxy-2-hydroxypropyl) acetate
Molecular Weight (g/mol) 176.17
Formula Weight 176.0685
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.