missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butylbenzimidazolium chloride, 96%, Thermo Scientific™

Product Code. p-4228579 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 11996651

Brand: Thermo Scientific Acros 435085000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 946607-10-9
Molecular Formula C15H23ClN2
Molecular Weight (g/mol) 266.81
InChI Key CMQDFXSMZLANNA-UHFFFAOYSA-M
Synonym 1,3-di-tert-butylbenzimidazolium chloride, 1,3-di-tert-butyl-1,3-benzodiazol-1-ium chloride, 1,3-di-tert-butyl-1h-benzimidazol-3-ium chloride, 1,3-di-tert-butyl-1h-benzo d imidazol-3-ium chloride
PubChem CID 16218265
IUPAC Name 1,3-ditert-butylbenzimidazol-3-ium;chloride
SMILES CC(C)(C)N1C=[N+](C2=CC=CC=C21)C(C)(C)C.[Cl-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 204.0°C
Color Tan to White
Elemental Analysis 64.15 to 70.90% C, 9.97 to 11.02% N
Infrared Spectrum Authentic
Assay Percent Range 96%
Quantity 500 mg
Formula Weight 266.81
Percent Purity 96%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 1, 3-Di-tert-butylbenzimidazolium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Di-tert-butylbenzimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.