missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butylbenzene, 99%

Product Code. 11489796
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489796 25 g 25g
11469796 1 g 1g
11479796 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489796 Supplier Thermo Scientific Alfa Aesar Supplier No. B20290.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1014-60-4 | C14H22 | 190.33 g/mol

1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1014-60-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00008830
InChI Key ILNDSSCEZZFNGE-UHFFFAOYSA-N
Synonym 1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
PubChem CID 136810
IUPAC Name 1,3-ditert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC=C1)C(C)(C)C

Specifications

Melting Point 10°C to 11°C
Density 0.859
Boiling Point 106°C to 107°C (18 mmHg)
Flash Point 83°C (183°F)
Refractive Index 1.488
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 190.33
Percent Purity 99%
Chemical Name or Material 1,3-Di-tert-butylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.