missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-BOC-2-(trifluoromethylsulfonyl)guanidine, 98%, Thermo Scientific Chemicals

Product Code. 10614945
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207857-15-6
Molecular Formula C12H20F3N3O6S
Molecular Weight (g/mol) 391.36
InChI Key GOQZIPJCBUYLIR-UHFFFAOYSA-N
Synonym 1,3-di-boc-2-trifluoromethylsulfonyl guanidine, goodman's reagent, n,n'-di-boc-n-triflylguanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethanesulfonyl guanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethylsulfonyl guanidine, 1,3-di-boc-2-trifluoromethanesulfonyl guanidine, tert-butyl n-n-2-methylpropan-2-yl oxycarbonyl-n'-trifluoromethylsulfonyl carbamimidoyl carbamate, tert-butyl n-tert-butoxycarbonyl amino trifluoromethanesulfonylimino methyl carbamate, tert-butyl n-tert-butoxycarbonyl-n'-trifluoromethyl sulphonyl carbamimidoyl carbamate
PubChem CID 3694570
IUPAC Name tert-butyl N-[N-[(2-methylpropan-2-yl)oxycarbonyl]-N'-(trifluoromethylsulfonyl)carbamimidoyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=NS(=O)(=O)C(F)(F)F)NC(=O)OC(C)(C)C

Specifications

Melting Point 111°C to 113°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Formula Weight 391.36
Percent Purity 98%
Quantity 1 g
Chemical Name or Material 1, 3-Di-BOC-2-(trifluoromethylsulfonyl)guanidine
compliance-icons
Product Identifier
  • 1,3-Di-BOC-2-(trifluoromethylsulfonyl)guanidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'