missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclopentanedione, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 1,3-Cyclopentanedione |
| CAS | 3859-41-4 |
| Molecular Formula | C5H6O2 |
| Quantity | 10 g |
| Synonym | 1,3-Cyclopentadione, NSC 364015 |
| InChI Formula | InChI=1S/C5H6O2/c6-4-1-2-5(7)3-4/h1-3H2 |
| SMILES | O=C1CCC(=O)C1 |
| IUPAC Name | cyclopentane-1,3-dione |
| Molecular Weight (g/mol) | 98.1 |
| Formula Weight | 98.0368 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction