missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclopentanedione, 99%

Product Code. 10104340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104340 5 g Glass bottle 5g
10391384 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10104340 Supplier Thermo Scientific Acros Supplier No. 175720050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3859-41-4 | C5H6O2 | 98.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3859-41-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00001405
InChI Key LOGSONSNCYTHPS-UHFFFAOYSA-N
PubChem CID 77466
ChEBI CHEBI:41456
IUPAC Name cyclopentane-1,3-dione
SMILES O=C1CCC(=O)C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 148°C to 153°C
Color Ochre to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C5H6(=O)2
Quantity 5 g
Synonym 1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione
Formula Weight 98.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Cyclopentanedione
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.