missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclopentanedione, 98%

Product Code. 11398375
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398375 5 g 5g
11388375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398375 Supplier Thermo Scientific Alfa Aesar Supplier No. L10336.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3859-41-4 | C5H6O2 | 98.10 g/mol

1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3859-41-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00001405
InChI Key LOGSONSNCYTHPS-UHFFFAOYSA-N
PubChem CID 77466
ChEBI CHEBI:41456
IUPAC Name cyclopentane-1,3-dione
SMILES O=C1CCC(=O)C1

Specifications

Melting Point 151°C to 153°C
Quantity 5 g
Beilstein 1362728
Synonym 1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione
Solubility Information Very soluble in water.
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material 1,3-Cyclopentanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.