missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclooctadiene, 97%

Product Code. 11348475
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11348475

Brand: Thermo Scientific Alfa Aesar L10516.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1700-10-3 | C8H12 | 108.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1700-10-3
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001751
InChI Key RRKODOZNUZCUBN-CCAGOZQPSA-N
Synonym 1,3-cyclooctadiene, cyclooctadiene, 1z,3z-cycloocta-1,3-diene, cis,cis-1,3-cyclooctadiene, cycloocta-1,3-diene, 1,3-cyclooctadiene, z,z, cyclooctadienes, 1.3-cyclooctadiene, 1e,3z-1,3-cyclooctadiene, 1,3-cod
PubChem CID 299882
IUPAC Name (1Z,3Z)-cycloocta-1,3-diene
SMILES C1CC\C=C/C=C\C1

Specifications

Melting Point -53°C
Density 0.869
Boiling Point 142°C to 144°C
Flash Point 24°C (75°F)
Odor Characteristic
Refractive Index 1.493
Quantity 5 g
UN Number UN2520
Beilstein 1901033
Formula Weight 108.18
Percent Purity 97%
Chemical Name or Material 1,3-Cyclooctadiene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.