missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanedione, 97%

Product Code. 10112570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10112570 100 g 100g
10530021 500 g 500g
10629833 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10112570 Supplier Thermo Scientific Acros Supplier No. 111171000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-02-9 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-02-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00001585
InChI Key HJSLFCCWAKVHIW-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
PubChem CID 10434
ChEBI CHEBI:17766
IUPAC Name cyclohexane-1,3-dione
SMILES C1CC(=O)CC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >101°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 1% max. Sodium chloride (Argentometry)
Packaging Plastic bottle
Linear Formula C6H8(=O)2
Quantity 100 g
Beilstein 07, 554
Fieser 01,178
Merck Index 15, 3196
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform,acetone and,boiling benzene,slightly soluble in ether,carbon disulfide and,petroleum ether
Formula Weight 112.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 3-Cyclohexanedione, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Cyclohexanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.