missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanedione, 97%, may cont. up to 1% NaCl

Product Code. p-7081172
Click to view available options
Quantity:
25 g
50 g
100 g
1000 g
Unit Size:
1000g
100g
25g
50g
This item is not returnable. View return policy

Product Code. 11484230

Brand: Thermo Scientific Alfa Aesar A11109.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-02-9 | C6H8O2 | 112.128 g/mol

It is used as a nitisinone intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a nitisinone intermediate.

Solubility
Soluble in water (155 g/L) at 20°C, alcohol, chloroform, acetone, boiling benzene, ether (slightly), carbon disulfide (slightly), and petr ether (slightly).

Notes
Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-02-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001585
InChI Key HJSLFCCWAKVHIW-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
PubChem CID 10434
ChEBI CHEBI:17766
IUPAC Name cyclohexane-1,3-dione
SMILES C1CC(=O)CC(=O)C1

Specifications

Melting Point 101°C to 105°C
Density 1.1
Quantity 25 g
Beilstein 385888
Sensitivity Light sensitive
Merck Index 14,3178
Solubility Information Soluble in water (155g/L) at 20°C,alcohol,chloroform,acetone,boiling benzene,ether (slightly),carbon disulfide (slightly),and petr ether (slightly).
Formula Weight 112.13
Percent Purity ≥97%
Concentration or Composition (by Analyte or Components) May contain up to 1% NaCl
Chemical Name or Material 1,3-Cyclohexanedione
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.