missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanediol, cis + trans, 98%

Product Code. 11336796
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336796 25 g 25g
11326796 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336796 Supplier Thermo Scientific Alfa Aesar Supplier No. L04110.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-01-8 | C6H12O2 | 116.16 g/mol

1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-01-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00039458
InChI Key RLMGYIOTPQVQJR-UHFFFAOYSA-N
Synonym 1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
PubChem CID 10433
IUPAC Name cyclohexane-1,3-diol
SMILES C1CC(CC(C1)O)O

Specifications

Melting Point ∼30°C
Boiling Point 246°C to 247°C
Quantity 25 g
Beilstein 2036654
Solubility Information Soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1,3-Cyclohexanediol, Cis + Trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.