missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexadiene, TRC

Product Code. 30356935
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356935 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356935 Supplier Toronto Research Chemicals Supplier No. TRCC98796525ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Cyclohexadiene
CAS 592-57-4
Molecular Formula C6H8
Quantity 25 mL
Synonym Dihydro-benzene, Cyclohexa-1,3-diene
InChI Formula InChI=1S/C6H8/c1-2-4-6-5-3-1/h1-4H,5-6H2
SMILES C1CC=CC=C1
IUPAC Name cyclohexa-1,3-diene
Molecular Weight (g/mol) 80.13
Formula Weight 80.0626
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.