missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexadiene, 96%, stabilized

Product Code. 10500021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10500021 100 mL Glass bottle 100mL
10365790 5 mL Glass bottle 5mL
10487480 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10500021 Supplier Thermo Scientific Acros Supplier No. 111101000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-57-4 | C6H8 | 80.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-57-4
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001532
InChI Key MGNZXYYWBUKAII-UHFFFAOYSA-N
Synonym 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
PubChem CID 11605
ChEBI CHEBI:37610
IUPAC Name cyclohexa-1,3-diene
SMILES C1CC=CC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -98.0°C
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 80.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.4760
Quantity 100 mL
Specific Gravity 0.86
Solubility Information Solubility in water: immiscible.
Stabilizer 0.1% BHT
Formula Weight 80.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,3-Cyclohexadiene, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.