missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexadiene, 96%, stab. with 0.1% BHT

Product Code. 11403997
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403997 5 g 5g
11413997 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403997 Supplier Thermo Scientific Alfa Aesar Supplier No. A19394.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-57-4 | C6H8 | 80.13 g/mol

1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.

Solubility
Slightly miscible with methanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-57-4
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001532
InChI Key MGNZXYYWBUKAII-UHFFFAOYSA-N
Synonym 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
PubChem CID 11605
ChEBI CHEBI:37610
IUPAC Name cyclohexa-1,3-diene
SMILES C1CC=CC=C1

Specifications

Melting Point -89°C
Density 0.847
Boiling Point 79°C to 81°C
Flash Point −8°C (−144°F)
Odor Unpleasant
Refractive Index 1.474
Quantity 5 g
UN Number UN3295
Beilstein 506024
Solubility Information Slightly miscible with methanol. Immiscible with water.
Formula Weight 80.13
Percent Purity 96%
Chemical Name or Material 1,3-Cyclohexadiene, stabilized with 0.1% BHT
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Cyclohexadiene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Cyclohexadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.