missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenyl)piperazine hydrochloride, 97%, Thermo Scientific™

Catalog Number 15436338
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
65369-76-8
C10H14Cl2N2
233.136
MFCD00039032
MHXPYWFZULXYHT-UHFFFAOYSA-N
1-3-chlorophenyl piperazine hydrochloride, m-cpp hydrochloride, 1-3-chlorophenyl piperazine monohydrochloride, unii-7im2gs7ilv, 1-3-chlorophenyl piperazinium chloride, piperazine, 1-3-chlorophenyl-, monohydrochloride, 1-m-chlorophenyl piperazine hcl, m-chlorophenylpiperazine hydrochloride, 7im2gs7ilv, 1-m-chlorophenyl piperazine hydrochloride
25681
1-(3-chlorophenyl)piperazine;hydrochloride
C1CN(CCN1)C2=CC(=CC=C2)Cl.Cl
This item is not returnable. View return policy
65369-76-8
C10H14Cl2N2
233.136
MFCD00039032
MHXPYWFZULXYHT-UHFFFAOYSA-N
1-3-chlorophenyl piperazine hydrochloride, m-cpp hydrochloride, 1-3-chlorophenyl piperazine monohydrochloride, unii-7im2gs7ilv, 1-3-chlorophenyl piperazinium chloride, piperazine, 1-3-chlorophenyl-, monohydrochloride, 1-m-chlorophenyl piperazine hcl, m-chlorophenylpiperazine hydrochloride, 7im2gs7ilv, 1-m-chlorophenyl piperazine hydrochloride
25681
1-(3-chlorophenyl)piperazine;hydrochloride
C1CN(CCN1)C2=CC(=CC=C2)Cl.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 210°C to 214°C (decomposition)
Color Brown
Quantity 5 g
Formula Weight 196.68 (Anhydrous)
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(3-Chlorophenyl)piperazine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'