missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenyl)ethanol, 97%

Product Code. 11391707
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391707 25 g 25g
11381707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391707 Supplier Thermo Scientific Alfa Aesar Supplier No. L04783.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6939-95-3 | C8H9ClO | 156.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6939-95-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00021864
InChI Key QYUQVBHGBPRDKN-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl ethanol, 3-chloro-alpha-methylbenzyl alcohol, 1-3-chlorophenyl ethan-1-ol, 1-3-chlorophenyl-1-ethanol, 3-chlorophenylethanol, acmc-1cap8, 1-m-chlorophenyl ethanol, 3'-chloro-1-phenylethanol, 1-3-chlorophenyl-ethanol, 1-3-chlorophenyl ethyl alcohol
PubChem CID 97779
IUPAC Name 1-(3-chlorophenyl)ethanol
SMILES CC(C1=CC(=CC=C1)Cl)O

Specifications

Density 1.173
Boiling Point 83°C to 85°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5445
Quantity 25 g
Beilstein 2042021
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 1-(3-Chlorophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.