missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloroallyl)-1,3,5,7-tetraazaadamantan-1-ium Chloride, TRC

Product Code. 30134327
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134327 1 g 1g
30010925 100 mg 100mg
30005397 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134327 Supplier Toronto Research Chemicals Supplier No. TRCC3642031G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4080-31-3
Molecular Formula C9H16Cl2N4
Molecular Weight (g/mol) 251.16
Synonym 1-(3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantane Chloride, Methenamine 3-Chloroallylochloride, 1-(3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantane Chloride, 1-(3-Chloro-2-propenyl)-3,5,7-triaza-1-azoniatricyclo[3.3.1.13,7]decane Chloride;1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, 3,5,7-triaza-1-azoniatricyclo[3.3.1.13,7]decane deriv., 1-(3-Chloro-2-propenyl)-3,5,7-triaza-1-azoniatricyclo[3.3.1.13,7]decane Chloride;1-(3-Chloroallyl)-3,5,7-triaza-1-azoniadamantane Chloride;1-(3-Chloroallyl)-3,5,7-triaza-1-azoniumadamantane Chloride, Dowco 184, Dowicide Q, Dowicil 100, Dowicil 75, N-(3-Chloroallyl)hexaminium Chloride, Quaternium 15
IUPAC Name 1-[(E)-3-chloroprop-2-enyl]-3,5,7-triaza-1-azoniatricyclo[3.3.1.13,7]decane;chloride
SMILES [Cl-].ClC=CC[N+]12CN3CN(CN(C3)C1)C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H16ClN4.ClH/c10-2-1-3-14-7-11-4-12(8-14)6-13(5-11)9-14;/h1-2H,3-9H2;1H/q+1;/p-1/b2-1+;
Formula Weight 250.0752
Percent Purity >95%
Chemical Name or Material 1-(3-Chloroallyl)-1,3,5,7-tetraazaadamantan-1-ium Chloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.