missing translation for 'onlineSavingsMsg'
Learn More

1-[3-chloro-5-(trifluoromethyl)-2-pyridyl]piperazine, 97%, Thermo Scientific™

£7.00 - £25.00

Chemical Identifiers

CAS 132834-59-4
Molecular Formula C10H11ClF3N3
Molecular Weight (g/mol) 265.66
MDL Number MFCD00173934
InChI Key RRIPRMGRQRYRRG-UHFFFAOYSA-N
Synonym 1-3-chloro-5-trifluoromethyl-2-pyridyl piperazine, 1-3-chloro-5-trifluoromethyl pyridin-2-yl piperazine, 1-3-chloro-5-trifluoromethyl pyrid-2-yl piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl piperazine, piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl, 3-chloro-5-trifluoromethyl-2-piperazinopyridine, 1-3-chloro-5-trifluoromethyl-pyridin-2-yl-piperazine, piperazine,1-3-chloro-5-trifluoromethyl-2-pyridinyl
PubChem CID 667619
IUPAC Name 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazine
SMILES FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCNCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10190652
View Documents Promotion Details
Thermo Scientific Maybridge
SEW04036CB
250 mg Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10753663
View Documents Promotion Details
Thermo Scientific Maybridge
SEW04036DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

132834-59-4
265.66
RRIPRMGRQRYRRG-UHFFFAOYSA-N
667619
FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCNCC1
C10H11ClF3N3
MFCD00173934
1-3-chloro-5-trifluoromethyl-2-pyridyl piperazine, 1-3-chloro-5-trifluoromethyl pyridin-2-yl piperazine, 1-3-chloro-5-trifluoromethyl pyrid-2-yl piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl piperazine, piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl, 3-chloro-5-trifluoromethyl-2-piperazinopyridine, 1-3-chloro-5-trifluoromethyl-pyridin-2-yl-piperazine, piperazine,1-3-chloro-5-trifluoromethyl-2-pyridinyl
1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazine

Specifications

132834-59-4
97%
MFCD00173934
RRIPRMGRQRYRRG-UHFFFAOYSA-N
1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazine
667619
97%
97.0
C10H11ClF3N3
1-3-chloro-5-trifluoromethyl-2-pyridyl piperazine, 1-3-chloro-5-trifluoromethyl pyridin-2-yl piperazine, 1-3-chloro-5-trifluoromethyl pyrid-2-yl piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl piperazine, piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl, 3-chloro-5-trifluoromethyl-2-piperazinopyridine, 1-3-chloro-5-trifluoromethyl-pyridin-2-yl-piperazine, piperazine,1-3-chloro-5-trifluoromethyl-2-pyridinyl
FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCNCC1
265.66
265.66
1-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]piperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications