missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloro-2-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10493172
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10493172 1 g Amber Glass Bottle 1g
10332383 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10493172 Supplier Thermo Scientific Maybridge Supplier No. BTB10071DA
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3226-34-4
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00100492
InChI Key GBWVDQBTXFIIJF-UHFFFAOYSA-N
Synonym 1-3-chloro-2-hydroxyphenyl ethanone, 1-3-chloro-2-hydroxyphenyl ethan-1-one, 3'-chloro-2'-hydroxyacetophenone, ethanone,1-3-chloro-2-hydroxyphenyl, 2'-hydroxy-3'-chloroacetophenone, 2'-hydroxy-3'-chloro acetophenone, 1-3-chloro-2-hydroxyphenyl-ethanone, 1-3-chloro-2-hydroxy-phenyl-ethanone, 1-3-chloro-2-hydroxyphenyl ethanone #, 2 inverted exclamation mark-hydroxy-3-chloroacetophenone
PubChem CID 593414
IUPAC Name 1-(3-chloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C(Cl)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.59
Percent Purity 97%
Grade Technical
Chemical Name or Material 1-(3-Chloro-2-hydroxyphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.