missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1,3-Butanediol, 99%

Product Code. 11499173
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499173 500 mL 500mL
11409183 2500 mL 2500mL
11419183 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499173 Supplier Thermo Scientific Alfa Aesar Supplier No. A11684.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-88-0 | C4H10O2 | 90.122 g/mol

(±)-1,3-Butanediol acts as a co-monomer in the production of polyurethane and polyester resins. It is used as a humectant (to prevent loss of moisture) in cosmetics, especially in hair sprays and setting lotions. It is used in surfactants, inks, solvents for natural and synthetic flavorings. It is involved in the synthesis of dual peroxisome proliferator-activated gamma and delta agonists acting as euglycemic agents, which is used in the treatment of diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-1,3-Butanediol acts as a co-monomer in the production of polyurethane and polyester resins. It is used as a humectant (to prevent loss of moisture) in cosmetics, especially in hair sprays and setting lotions. It is used in surfactants, inks, solvents for natural and synthetic flavorings. It is involved in the synthesis of dual peroxisome proliferator-activated gamma and delta agonists acting as euglycemic agents, which is used in the treatment of diabetes.

Solubility
Miscible with water.

Notes
Hygroscopic. Air and moisture sensitive. Incompatible with strong oxidizing agents, acid chlorides, chloroformates and reducing agents. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-88-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00004554
InChI Key PUPZLCDOIYMWBV-UHFFFAOYSA-N
Synonym 1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol
PubChem CID 7896
ChEBI CHEBI:52683
IUPAC Name butane-1,3-diol
SMILES CC(CCO)O

Specifications

Melting Point -50°C
Density 1.005
Boiling Point 203°C to 204°C
Flash Point 109°C (249°F)
Odor Mild
Refractive Index 1.44
Quantity 500 mL
Beilstein 1731276
Sensitivity Hygroscopic
Merck Index 14,1567
Solubility Information Miscible with water.
Formula Weight 90.12
Percent Purity 99%
Chemical Name or Material (±)-1,3-Butanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.