missing translation for 'onlineSavingsMsg'
Learn More

1,3-Butadiene diepoxide, 98+%, AcroSeal™, Thermo Scientific™

Product Code. 11954761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11954761 50 mL AcroSeal™ Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11954761 Supplier Thermo Scientific Acros Supplier No. 330070500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1464-53-5
CAS Min % 98.0
CAS Max % 100.0
Melting Point 2.0°C to 4.0°C
Density 1.1100g/mL
Boiling Point 56.0°C to 58.0°C (25.0 mmHg)
Flash Point 45°C
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C4H6O2
Refractive Index 1.4325 to 1.4345
Quantity 50 mL
Specific Gravity 1.11
Synonym diepoxybutane, 1,3-butadiene diepoxide, 2,2'-bioxirane, butadiene diepoxide, dioxybutadiene, butadiene dioxide, bioxirane, butane diepoxide, 1,2:3,4-diepoxybutane, butadiendioxyd
Solubility Information Solubility in water: miscible with water, hydrolyzes. Other solubilities: miscible with ethanol, react with alcohols, acids, amines, soluble in aprotonic organic solvents
InChI Key ZFIVKAOQEXOYFY-UHFFFAOYSA-N
SMILES C1C(O1)C2CO2
IUPAC Name 2-(oxiran-2-yl)oxirane
Molecular Weight (g/mol) 86.09
PubChem CID 11254
ChEBI CHEBI:23704
Formula Weight 86.09
Percent Purity 98+%
Chemical Name or Material 1,3-Butadiene diepoxide, 98+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.