missing translation for 'onlineSavingsMsg'
Learn More

1,3-Butadiene 400 μg/mL in N,N-Dimethylacetamide, Dr. Ehrenstorfer

Product Code. 30317211
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30317211

Brand: Dr. Ehrenstorfer DREV10860500DA400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Butadiene
CAS 106-99-0
Analyte or Component Names 1,3-Butadiene
Shipping Condition Room Temperature
Molecular Formula C4 H6
Concentration 400 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,3-Butadiene, Biethylene, Bivinyl, Butadiene, Butadiene-1,3, Divinyl, Erythrene, Vinylethylene, α,γ-Butadiene
InChI Formula InChI=1S/C4H6/c1-3-4-2/h3-4H,1-2H2
SMILES C=CC=C
IUPAC Name buta-1,3-diene
Solvent or Matrix N,N-Dimethylacetamide
Industry Type Food Testing
Molecular Weight (g/mol) 54.09
Quantity 5 mL
Formula Weight 54.047 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.