missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromopropoxy)-3-chlorobenzene, 96%, Thermo Scientific™

Product Code. 10220313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10220313 25 g Glass bottle 25g
10506553 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10220313 Supplier Thermo Scientific Acros Supplier No. 379230250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 37142-46-4
Molecular Formula C9H10BrClO
Molecular Weight (g/mol) 249.53
MDL Number MFCD00596672
InChI Key MVKLHPFBHSJVLA-UHFFFAOYSA-N
Synonym 1-3-bromopropoxy-3-chlorobenzene, 3-bromopropyl 3-chlorophenyl ether
PubChem CID 2734929
IUPAC Name 1-(3-bromopropoxy)-3-chlorobenzene
SMILES C1=CC(=CC(=C1)Cl)OCCCBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Boiling Point 139°C to 148°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4O(CH2)3Br
Refractive Index 1.5570 to 1.559
Quantity 25 g
Formula Weight 249.53
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-(3-Bromopropoxy)-3-chlorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(3-Bromopropoxy)-3-chlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.