missing translation for 'onlineSavingsMsg'
Learn More

1-(3-bromopropoxy)-2-nitrobenzene, 90+%, Thermo Scientific™

Product Code. p-3211213 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10341263

Brand: Thermo Scientific Acros 385240050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 104147-69-5
MDL Number MFCD00596660
InChI Key HPZBIRIHQKSGGT-UHFFFAOYSA-N
Synonym 1-3-bromopropoxy-2-nitrobenzene, 1-3-bromo-propoxy-2-nitro-benzene
PubChem CID 7172300
IUPAC Name 1-(3-bromopropoxy)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 36°C to 39°C
Color Yellow
Boiling Point 131°C to 132°C (0.5 mmHg)
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Molecular Formula C9H10BrNO3
Linear Formula NO2C6H4O(CH2)3Br
Quantity 5 g
Molecular Weight (g/mol) 260.09
Formula Weight 260.09
Percent Purity 90+%
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Bromopropoxy)-2-nitrobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(3-Bromopropoxy)-2-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.