missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(trimethylsilyl)urea, TRC

Product Code. 30331869
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30331869 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30331869

Brand: Toronto Research Chemicals TRCB535405500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Bis(trimethylsilyl)urea
CAS 18297-63-7
Molecular Formula C7 H20 N2 O Si2
Quantity 500 mg
Synonym Bis(trimethylsilyl)urea, Bis-TMS urea
InChI Formula InChI=1S/C7H20N2OSi2/c1-11(2,3)8-7(10)9-12(4,5)6/h1-6H3,(H2,8,9,10)
SMILES C[Si](C)(C)NC(=O)N[Si](C)(C)C
IUPAC Name 1,3-bis(trimethylsilyl)urea
Molecular Weight (g/mol) 204.42
Formula Weight 204.1114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.