missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(trifluoromethyl)benzene, 98+%

Product Code. p-7081345
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11450163

Brand: Thermo Scientific Alfa Aesar A11360.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 402-31-3 | C8H4F6 | 214.11 g/mol

1,3-Bis(trifluoromethyl)benzene undergoes regioselective metalation and subsequent carboxylation at position 2 to give 2,6-bis(trifluoromethyl)benzoic acid. a convenient, selective synthesis of bis[2,4-bis(trifluoromethyl)phenyl]phosphane derivatives with 1,3-bis(trifluoromethyl)benzene as starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(trifluoromethyl)benzene undergoes regioselective metalation and subsequent carboxylation at position 2 to give 2,6-bis(trifluoromethyl)benzoic acid. a convenient, selective synthesis of bis[2,4-bis(trifluoromethyl)phenyl]phosphane derivatives with 1,3-bis(trifluoromethyl)benzene as starting material.

Solubility
Insoluble in water. Soluble in alcohol, ether, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-31-3
Molecular Formula C8H4F6
Molecular Weight (g/mol) 214.11
MDL Number MFCD00000392
InChI Key SJBBXFLOLUTGCW-UHFFFAOYSA-N
Synonym 1,3-bis trifluoromethyl benzene, benzene, 1,3-bis trifluoromethyl, 1,3-di trifluoromethyl benzene, 1,3-bis trifluoromethyl-benzene, m-bis trifluoromethyl benzene, m-ditrifluoromethylbenzene, hexafluoro-m-xylene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-m-xylene, mtf-tfm, a,a,a,a',a',a'-hexafluoro-m-xylene
PubChem CID 223106
IUPAC Name 1,3-bis(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point -35°C
Density 1.378
Boiling Point 115°C to 117°C
Flash Point 26°C (78°F)
Odor Aromatic
Refractive Index 1.379
Quantity 50 g
UN Number UN1993
Beilstein 2052589
Solubility Information Insoluble in water. Soluble in alcohol,ether,benzene.
Formula Weight 214.11
Percent Purity ≥98%
Chemical Name or Material 1,3-Bis (trifluoromethyl)benzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.