missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(methylthio)propane, 97%

Product Code. 11420517
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420517 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420517 Supplier Thermo Scientific Alfa Aesar Supplier No. B22397.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24949-35-7 | C5H12S2 | 136.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24949-35-7
Molecular Formula C5H12S2
Molecular Weight (g/mol) 136.27
MDL Number MFCD00039811
InChI Key JIZDZCFZPMIBPK-UHFFFAOYSA-N
Synonym 1,3-bis methylthio propane, 1,3-bis methylsulfanyl propane, 2,6-dithiaheptane, propane, 1,3-bis methylthio, acmc-20ap52, 1,3-bis methylsulfanyl-propane
PubChem CID 141161
IUPAC Name 1,3-bis(methylsulfanyl)propane
SMILES CSCCCSC

Specifications

Density 1.011
Boiling Point 88°C to 90°C (12 mmHg)
Flash Point >100°C (212°F)
Odor Stench
Refractive Index 1.519
Quantity 5 g
Beilstein 1732333
Formula Weight 136.28
Percent Purity 97%
Chemical Name or Material 1,3-Bis(methylthio)propane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.