missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(diphenylphosphino)propane, 97%

Product Code. 10227223 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10227223

Brand: Thermo Scientific Acros 310050250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6737-42-4 | C27H26P2 | 412.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6737-42-4
Molecular Formula C27H26P2
Molecular Weight (g/mol) 412.45
MDL Number MFCD00003050
InChI Key LVEYOSJUKRVCCF-UHFFFAOYSA-N
Synonym 1,3-bis diphenylphosphino propane, dppp, trimethylenebis diphenylphosphine, phosphine, 1,3-propanediylbis diphenyl, 3-diphenylphosphanyl propyl diphenylphosphane, 3-diphenylphosphanylpropyl diphenyl phosphane, 1,3-bis diphenyphosphino propane, 1,3-bis diphenylphosphino propane dppp, bis 1,3-diphenylphosphino propane, 1,3-bis-diphenylphosphino propane
PubChem CID 81219
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57.0°C to 63.0°C
Elemental Analysis 96.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)2P(CH2)3P(C6H5)2
Quantity 25 g
Merck Index 15,1254
Solubility Information Solubility in water: insoluble
IUPAC Name [3-(diphenylphosphanyl)propyl]diphenylphosphane
Formula Weight 412.46
Percent Purity 97%
Physical Form Powder or Granules
Chemical Name or Material 1, 3-Bis(diphenylphosphino)propane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.