missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-piperidinyl)propane, 97+%

Product Code. 11395679
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11395679

Brand: Thermo Scientific Alfa Aesar L20442.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16898-52-5 | C13H26N2 | 210.365 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16898-52-5
Molecular Formula C13H26N2
Molecular Weight (g/mol) 210.365
MDL Number MFCD00038013
InChI Key OXEZLYIDQPBCBB-UHFFFAOYSA-N
Synonym 1,3-bis 4-piperidyl propane, 4,4'-trimethylenedipiperidine, 1,3-di-4-piperidylpropane, piperidine, 4,4'-1,3-propanediyl bis, di-pip, 4-3-piperidin-4-yl propyl piperidine, 4-3-4-piperidyl propyl piperidine, unii-eu53ec219i, 1,3-di 4-piperidyl propane, 1,3-bis 4-piperidinyl propane
PubChem CID 85631
IUPAC Name 4-(3-piperidin-4-ylpropyl)piperidine
SMILES C1CNCCC1CCCC2CCNCC2

Specifications

Melting Point 63°C to 68°C
Boiling Point 327°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 111615
Sensitivity Hygroscopic
Formula Weight 210.36
Percent Purity ≥97%
Chemical Name or Material 1,3-Bis(4-piperidinyl)propane
compliance-icons compliance-icons
Product Identifier
  • 1,3-Bis(4-piperidinyl)propane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Bis(4-piperidinyl)propane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.