missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate, 98%, Thermo Scientific™

Product Code. 15425785
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425785 25 g 25g
15435785 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425785 Supplier Thermo Scientific Alfa Aesar Supplier No. H66513.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915967-82-7
Molecular Formula C29H52Cl2N6O20P4
Molecular Weight (g/mol) 999.552
MDL Number MFCD11870901
InChI Key AMCQDGFOKTXHSY-UHFFFAOYSA-N
Synonym piperaquine phosphate hydrate, piperaquine tetraphosphate tetrahydrate, unii-854d7k8lxb, piperaquine tetraphosphate hydrate, piperaquine phosphate, quinoline, 4,4'-1,3-propanediyldi-4,1-piperazinediyl bis 7-chloro-, phosphate, hydrate 1:4:4, piperaquine tetraphosphate tetrahydrate hplc, tetrakis phosphoric acid piperaquine tetrahydrate, 1,3-bis 4-7-chloroquinolin-4-yl piperazin-1-yl propane tetrakis phosphate tetrahydrate
PubChem CID 49849842
IUPAC Name 7-chloro-4-[4-[3-[4-(7-chloroquinolin-4-yl)piperazin-1-yl]propyl]piperazin-1-yl]quinoline;phosphoric acid;tetrahydrate
SMILES C1CN(CCN1CCCN2CCN(CC2)C3=C4C=CC(=CC4=NC=C3)Cl)C5=C6C=CC(=CC6=NC=C5)Cl.O.O.O.O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O

Specifications

Color White
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 999.55
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.