missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene, 96%

Product Code. p-7046946
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11333826

Brand: Thermo Scientific Alfa Aesar L16894.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 802-93-7 | C12H6F12O2 | 410.159 g/mol

1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene is used to determine the specific details of electrical magnitudes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene is used to determine the specific details of electrical magnitudes.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 802-93-7
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.159
MDL Number MFCD00042090
InChI Key PGUIOHNOYADLMU-UHFFFAOYSA-N
Synonym 1,3-bis 2-hydroxyhexafluoroisopropyl benzene, 1,3-bis hexafluoro-2-hydroxyisopropyl benzene, 1,3-bis alpha-hydroxyhexafluoroisopropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,3-benzenedimethanol, a,a,a',a'-tetrakis trifluoromethyl-1,3-benzenedimethanol, 1,3-benzenedimethanol, .alpha.,.alpha.,.alpha.',.alpha.'-tetrakis trifluoromethyl, 1,3-bis hexafluoro-alpha-hydroxyisopropyl benzene, 1,1,1,3,3,3-hexafluoro-2-3-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,3-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, 1,3 hfab
PubChem CID 69933
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES C1=CC(=CC(=C1)C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O

Specifications

Melting Point 9°C to 10°C
Density 1.659
Boiling Point 99°C to 100°C (20 mmHg)
Flash Point 97°C (206°F)
Refractive Index 1.389
Quantity 5 g
Beilstein 2024879
Solubility Information Difficult to mix.
Formula Weight 410.16
Percent Purity 96%
Chemical Name or Material 1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.