missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazol-6-amine, 97+%, Thermo Scientific™

Product Code. 10650325
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 533-30-2
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.199
InChI Key FAYAYUOZWYJNBD-UHFFFAOYSA-N
Synonym 6-aminobenzothiazole, 6-benzothiazolamine, benzo d thiazol-6-amine, 6-amino-benzothiazole, benzothiazole, 6-amino, benzothiazol-6-amine, benzothiazol-6-ylamine, unii-9yfg4wuj1g, 9yfg4wuj1g, 1,3-benzothiazol-6-ylamine
PubChem CID 68288
IUPAC Name 1,3-benzothiazol-6-amine
SMILES C1=CC2=C(C=C1N)SC=N2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Merck Index 15, 420
Formula Weight 150.2
Percent Purity 97%
Chemical Name or Material 1, 3-Benzothiazol-6-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'