missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazol-5-amine, 97%, Thermo Scientific™

Product Code. 10397114
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397114 10 g Amber Glass Bottle 10g
10218862 250 mg Amber Glass Bottle 250mg
10530093 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10397114 Supplier Thermo Scientific Maybridge Supplier No. MO07036EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1123-93-9
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.20
MDL Number MFCD04115282
InChI Key UJZYHMZRXGNDFB-UHFFFAOYSA-N
Synonym 5-benzothiazolamine, benzothiazol-5-ylamine, benzo d thiazol-5-amine, benzothiazol-5-amine, 5-amino-1,3-benzothiazole, 5-aminobenzothiazole, 1,3-benzothiazole-5-amine, 1,3-benzothiazol-5-ylamine, 5-amino-1,3-benzothiazol, 5-amino-benzothiazole
PubChem CID 70749
IUPAC Name 1,3-benzothiazol-5-amine
SMILES NC1=CC=C2SC=NC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 150.2
Percent Purity 95%
Chemical Name or Material 1, 3-Benzothiazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.