missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxol-5-ylmethyl isothiocyanate, ≥95%, Thermo Scientific™

Catalog Number 10273752
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
4430-47-1
C9H7NO2S
193.22
MFCD00041217
PUJWRDBPAFJUJW-UHFFFAOYSA-N
3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798
2795371
5-(isothiocyanatomethyl)-1,3-benzodioxole
S=C=NCC1=CC=C2OCOC2=C1
This item is not returnable. View return policy
4430-47-1
C9H7NO2S
193.22
MFCD00041217
PUJWRDBPAFJUJW-UHFFFAOYSA-N
3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798
2795371
5-(isothiocyanatomethyl)-1,3-benzodioxole
S=C=NCC1=CC=C2OCOC2=C1
CAS Min % 95.0
Density 1.32g/mL
Assay Percent Range 95%
Quantity 1 g
Formula Weight 193.23
Percent Purity 95%
Chemical Name or Material 1, 3-Benzodioxol-5-ylmethyl isothiocyanate
compliance-icons
Product Identifier
  • 1,3-Benzodioxol-5-ylmethyl isothiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'