missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxol-5-ylmethyl isothiocyanate, ≥95%, Thermo Scientific™

Product Code. 11326296
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326296 10 g 10g
10273752 1 g 1g
10556615 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326296 Supplier Thermo Scientific Maybridge Supplier No. S03337EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4430-47-1
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041217
InChI Key PUJWRDBPAFJUJW-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798
PubChem CID 2795371
IUPAC Name 5-(isothiocyanatomethyl)-1,3-benzodioxole
SMILES S=C=NCC1=CC=C2OCOC2=C1

Specifications

CAS Min % 95.0
Density 1.32g/mL
Assay Percent Range 95%
Quantity 10 g
Formula Weight 193.23
Percent Purity 95%
Chemical Name or Material 1, 3-Benzodioxol-5-ylmethyl isothiocyanate
compliance-icons
Product Identifier
  • 1,3-Benzodioxol-5-ylmethyl isothiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.