missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedithiol, 95%

Product Code. 15555186 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15555186 5 g 5g
15545186 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15555186 Supplier Thermo Scientific Acros Supplier No. 456100050
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626-04-0 | C6H6S2 | 142.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-04-0
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004842
InChI Key ZWOASCVFHSYHOB-UHFFFAOYSA-N
Synonym 1,3-benzenedithiol, 1,3-dimercaptobenzene, dithioresorcin, dithioresorcinol, 1,3-benzedithiol, acmc-20ap5g, ksc357g0j
PubChem CID 522062
IUPAC Name benzene-1,3-dithiol
SMILES SC1=CC(S)=CC=C1

Specifications

Melting Point 24.0°C
Density 1.2400g/mL
Boiling Point 128°C to 129°C (21.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Linear Formula C6H4(SH)2
Quantity 5 g
Specific Gravity 1.24
Formula Weight 142.24
Percent Purity 95%
Chemical Name or Material 1,3-Benzenedithiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Benzenedithiol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.