missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedisulfonyl chloride, 98%, Thermo Scientific™

Product Code. 10751632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10751632 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10751632 Supplier Thermo Scientific Acros Supplier No. 401810250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 585-47-7
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H4Cl2O4S2
Linear Formula C6H4(SO2Cl)2
MDL Number MFCD00041879
Quantity 25g
Synonym 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
Solubility Information Solubility in water: reacts
InChI Key ALIQZUMMPOYCIS-UHFFFAOYSA-N
SMILES ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O
IUPAC Name benzene-1,3-disulfonyl dichloride
Molecular Weight (g/mol) 275.11
PubChem CID 96240
Formula Weight 275.13
Percent Purity 98%
Chemical Name or Material 1, 3-Benzenedisulfonyl chloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.