missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedisulfonyl chloride, 97%

Product Code. 11466956
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466956 5 g 5g
11476956 25 g 25g
11486956 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466956 Supplier Thermo Scientific Alfa Aesar Supplier No. B24044.06
  • ONLINE EXCLUSIVE PRICE
    £38.10
    £34.65 / 5g
    Save £3.45
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 585-47-7 | C6H4Cl2O4S2 | 275.11 g/mol

1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

Solubility
Soluble in DMSO and methanol. Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-47-7
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.11
MDL Number MFCD00041879
InChI Key ALIQZUMMPOYCIS-UHFFFAOYSA-N
Synonym 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
PubChem CID 96240
SMILES ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 58°C to 60°C
Quantity 5 g
UN Number UN3261
Beilstein 1118508
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO and methanol. Reacts with water.
IUPAC Name benzene-1,3-disulfonyl dichloride
Formula Weight 275.13
Percent Purity 98%
Chemical Name or Material 1,3-Benzenedisulfonyl chloride
compliance-icons
Product Identifier
  • 1,3-Benzenedisulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Benzenedisulfonyl dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.