missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Benzenedimethanol, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 56.0°C to 60.0°C |
| Color | Colorless to Yellow |
| Boiling Point | 154.0°C to 159.0°C (13.0 mmHg) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H4(CH2OH)2 |
| Quantity | 2 g |
| Show More |
Safety and Handling
Product Identifier
- 1,3-Benzenedimethanol
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur