missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedimethanamine, TRC

Product Code. 30136913
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136913 500 mg 500mg
30136397 2.5 g 2.5g
17975055 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136913 Supplier Toronto Research Chemicals Supplier No. TRCB203578500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-55-0
Formula Weight 136.1
InChI Formula InChI=1S/C8H12N2/c9-5-7-2-1-3-8(4-7)6-10/h1-4H,5-6,9-10H2
IUPAC Name [3-(aminomethyl)phenyl]methanamine
Molecular Formula C8 H12 N2
Molecular Weight (g/mol) 136.19
SMILES NCc1cccc(CN)c1
Synonym m-Xylene-α,α'-diamine (6CI,8CI), 1,3-Benzenebis(methylamine), 1,3-Bis(aminomethyl)benzene, 1,3-Xylylenediamine, 3-(Aminomethyl)benzylamine, Aradur 22, Aramine 31-487, Benzene-1,3-diyldimethanamine, Epilink MX, Euredur 22, MXDA, NSC 61568, Shoamine X, m-(α,α'-Diamino)xylene, m-Xylene diamine, m-Xylylene-α,α'-diamine, m-Xylylenediamine, α,α'-Diamino-m-xylene, α,α'-m-Xylenediamine, 1,3-Benzenedimethanamine
Chemical Name or Material 1,3-Benzenebis(methylamine)

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.