missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminopropyl)pyrrolidine, 97%

Product Code. 11331697
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331697 25 g 25g
11321697 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331697 Supplier Thermo Scientific Alfa Aesar Supplier No. L04739.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23159-07-1 | C7H16N2 | 128.219 g/mol

1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .

Solubility
Soluble in water, soluble in chloroform

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23159-07-1
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00014100
InChI Key VPBWZBGZWHDNKL-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-yl propan-1-amine, 1-3-aminopropyl pyrrolidine, 1-3-aminopropyl-pyrrolidine, 1-pyrrolidinepropanamine, n-3-aminopropyl pyrrolidine, 3-pyrrolidinopropylamine, 3-1-pyrrolidinyl propylamine, pyrrolidine, 1-3-aminopropyl, 3-pyrrolidin-1-ylpropylamine, 3-1-pyrrolidino propylamine
PubChem CID 31670
IUPAC Name 3-pyrrolidin-1-ylpropan-1-amine
SMILES C1CCN(C1)CCCN

Specifications

Density 1.014
Boiling Point 191°C to 193°C
Refractive Index 1.471
Quantity 25 g
UN Number UN2735
Beilstein 1191
Sensitivity Air Sensitive
Solubility Information Soluble in water,soluble in chloroform
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material 1-(3-Aminopropyl)pyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.