missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminopropyl)imidazole, 98%

Product Code. p-7021716
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11432217

Brand: Thermo Scientific Alfa Aesar A14169.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5036-48-6 | C6H11N3 | 125.18 g/mol

1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents and air. Air sensitive. Store under dry inert gas. Incompatible with air and carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5036-48-6
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.18
MDL Number MFCD00009819
InChI Key KDHWOCLBMVSZPG-UHFFFAOYSA-N
Synonym 1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
PubChem CID 78736
SMILES NCCCN1C=CN=C1

Specifications

Melting Point -68°C
Density 1.062
pH 11.3
Boiling Point 296°C
Flash Point 154°C (309°F)
Odor Amine-like
Refractive Index 1.519
Quantity 50 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
IUPAC Name 3-(1H-imidazol-1-yl)propan-1-amine
Formula Weight 125.18
Percent Purity 98%
Chemical Name or Material 1-(3-Aminopropyl)imidazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(3-Aminopropyl)imidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1H-Imidazole-1-propanamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.