missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminopropyl)-4-methylpiperazine, 98%

Product Code. 11331737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331737 1 g 1g
11341737 5 g 5g
11351737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331737 Supplier Thermo Scientific Alfa Aesar Supplier No. L04876.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4572-03-6 | C8H19N3 | 157.26 g/mol

1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

Notes
Air sensitive. Keep in a dry, cool and well-ventilated place. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4572-03-6
Molecular Formula C8H19N3
Molecular Weight (g/mol) 157.26
MDL Number MFCD00014616
InChI Key RGUABPVONIGVAT-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl propan-1-amine, 1-3-aminopropyl-4-methylpiperazine, 1-piperazinepropanamine, 4-methyl, 3-4-methyl-1-piperazinyl propylamine, 4-3-aminopropyl-1-methylpiperazine, 1-methyl-4-3'-aminopropyl piperazine, 4-methyl-1-piperazinepropanamine, 3-4-methyl-piperazin-1-yl-propylamine, 3-4-methylpiperazin-1-yl propylamine, n-methyl-n'-3-aminopropyl piperazine
PubChem CID 78328
IUPAC Name 3-(4-methylpiperazin-1-yl)propan-1-amine
SMILES CN1CCN(CC1)CCCN

Specifications

Density 0.924
Boiling Point 113°C to 114°C (13 mmHg)
Refractive Index 1.482
Quantity 1 g
UN Number UN2735
Beilstein 105964
Sensitivity Air Sensitive
Formula Weight 157.26
Percent Purity 98%
Chemical Name or Material 1-(3-Aminopropyl)-4-methylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.