missing translation for 'onlineSavingsMsg'
Learn More

1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one, 97%, Thermo Scientific™

Product Code. 10643455
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10643455 10 g 10g
10089251 250 mg 250mg
10456082 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10643455 Supplier Thermo Scientific Maybridge Supplier No. GK00604EA
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-02-7
Molecular Formula C12H10ClNOS
Molecular Weight (g/mol) 251.73
MDL Number MFCD00068162
InChI Key DLQVOIFYJQATMI-UHFFFAOYSA-N
Synonym 1-3-amino-5-4-chlorophenyl-2-thienyl ethan-1-one, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethanone, 2-acetyl-3-amino-5-4-chlorophenyl thiophene, ethanone,1-3-amino-5-4-chlorophenyl-2-thienyl, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethan-1-one
PubChem CID 2777494
SMILES CC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name 1-[3-amino-5-(4-chlorophenyl)thiophen-2-yl]ethanone
Formula Weight 251.74
Percent Purity 97%
Chemical Name or Material 1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.