missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Allyl-4-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10510942
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10510942 1 g Amber Glass Bottle 1g
10344532 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10510942 Supplier Thermo Scientific Maybridge Supplier No. KM05715DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1132-05-4
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00068015
InChI Key XVTCWUFLNLZPEJ-UHFFFAOYSA-N
PubChem CID 259653
IUPAC Name 1-(4-hydroxy-3-prop-2-enylphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)CC=C

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3'-allyl-4'-hydroxyacetophenone, acetophenone, 3'-allyl-4'-hydroxy, 1-3-allyl-4-hydroxyphenyl ethan-1-one, 1-3-allyl-4-hydroxyphenyl ethanone, ethanone, 1-4-hydroxy-3-2-propenyl phenyl, 1-4-hydroxy-3-prop-2-en-1-yl phenyl ethanone, ethanone,1-4-hydroxy-3-2-propen-1-yl phenyl, ethanone, 1-4-hydroxy-3-2-propen-1-yl phenyl, zlchem 1234
Formula Weight 176.21
Percent Purity 95%
Chemical Name or Material 1-(3-Allyl-4-hydroxyphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.